Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM62 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116643 Supplier Invitrogen™ Supplier No. PA5116643
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-431, Mouse brain Immunogen sequence: HEQHQVTGID DAFDELQREL KDQLQALQDS EREHTEALQL LKRQLAETKS STKSLRTTIG EAFERLHRLL RERQKAMLEE LEADTARTLT DIEQKVQRYS QQLRKVQEGA QILQERLAET DRHTFLAGVA SLSERLKGKI HETNLTYEDF PTSKYTGPLQ Y.

E3 ubiquitin ligase whose activity is dependent on E2 ubiquitin-conjugating enzyme UBE2D2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM62
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TRIM62
Gene Accession No. Q80V85, Q9BVG3
Gene Alias 6330414G21Rik; AI450348; Dear1; ductal epithelium-associated RING Chromosome 1; E3 ubiquitin-protein ligase TRIM62; RING-type E3 ubiquitin transferase TRIM62; TRIM62; tripartite motif containing 62; tripartite motif-containing 62; tripartite motif-containing protein 62
Gene Symbols TRIM62
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-280 of human TRIM62 (NP_0606772).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313045, 55223, 67525
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.