Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM64 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546652 Supplier Invitrogen™ Supplier No. PA546652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KPNFNTNVVL KKLSSLARQT RPQNINSSDN ICVLHEETKE LFCEADKRLL Sequence homology: Human: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM64
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIM64
Gene Accession No. A6NGJ6
Gene Alias C11orf28; TRIM64; TRIM64A; tripartite motif containing 64; tripartite motif-containing protein 64
Gene Symbols TRIM64
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TRIM64 (aa 61-110).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120146
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.