Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM65 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598519 Supplier Invitrogen™ Supplier No. PA598519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRIM65 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

TRIM65 is a protein coding gene and gene ontology (GO) includes cytosol, nucleoplasm, nucleus, positive regulation of autophagy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM65
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TRIM65
Gene Accession No. Q6PJ69
Gene Alias 4732463G12Rik; TRIM65; tripartite motif containing 65; tripartite motif-containing protein 65
Gene Symbols TRIM65
Host Species Rabbit
Immunogen Recombinant Human Tripartite motif-containing protein 65 protein (290-517AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201292
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.