Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM66 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569788
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569788 Supplier Invitrogen™ Supplier No. PA569788
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 85%; Horse: 86%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 79%.

TRIM66 gene ontology annotations related to this gene include zinc ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM66
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIM66
Gene Accession No. O15016
Gene Alias C11orf29; KIAA0298; Kiaa0298-hp; Tif1d; TIF1DELTA; transcriptional intermediary factor 1 delta; Trim66; tripartite motif containing 66; tripartite motif-containing 66; tripartite motif-containing protein 66
Gene Symbols TRIM66
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human TRIM66.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9866
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.