Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM67 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542274
Description
Peptide sequence: GGVCKGATVG VLLDLNKHTL TFFINGQQQG PTAFSHVDGV FMPALSLNRN Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 92%; Horse: 92%; Human: 92%; Mouse: 92%; Rabbit: 92%; Rat: 92%.
The TRIM67 gene encodes a member of the tripartite motif (TRIM) protein family, which is involved in various cellular processes including protein ubiquitination, signal transduction, and immune responses. TRIM67 contains RING, B-box, and coiled-coil domains, characteristic of TRIM proteins, which facilitate interactions with other proteins and contribute to its role in ubiquitination and cellular regulation. TRIM67 is notably expressed in the brain and has been associated with neurodevelopmental processes, particularly influencing neuronal differentiation and synapse formation. This gene plays a regulatory role in signaling pathways that are critical for neural development, and mutations or dysregulation in TRIM67 have been linked to neurodevelopmental disorders and cognitive deficits. Additionally, TRIM67's involvement in protein degradation and signal modulation may impact cellular architecture and brain function. Ongoing research seeks to elucidate TRIM67's precise mechanisms and potential therapeutic targets for neurological conditions.
Specifications
TRIM67 | |
Polyclonal | |
Unconjugated | |
TRIM67 | |
D130049O21Rik; RGD1560667; TNL; Trim67; TRIM9-like protein; TRIM9-like protein TNL; tripartite motif containing 67; tripartite motif-containing 67; tripartite motif-containing protein 67 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
440730 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6ZTA4 | |
TRIM67 | |
Synthetic peptide directed towards the C-terminal of human TRIM67 (aa 711-760). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction