Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM68 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540893
Description
Peptide sequence: RLRKGNEYRA GTDEYPILSL PVPPRRVGIF VDYEAHDISF YNVTDCGSHI Sequence homology: Cow: 93%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 93%; Rat: 86%.
The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is expressed in many cancer cell lines. Its expression in normal tissues, however, was found to be restricted to prostate. This gene was also found to be differentially expressed in androgen-dependent versus androgen-independent prostate cancer cells.
Specifications
TRIM68 | |
Polyclonal | |
Unconjugated | |
TRIM68 | |
AI894158; E3 ubiquitin-protein ligase TRIM68; F730114J12Rik; FLJ10369; GC109; LOC100720935; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM68; MGC126176; ring finger protein 137; RING-type E3 ubiquitin transferase TRIM68; RNF137; Ro/SSA1 related protein; SS56; SS-56; SSA protein SS-56; Trim68; tripartite motif containing 68; tripartite motif-containing 68; tripartite motif-containing protein 68 | |
Rabbit | |
Protein A | |
RUO | |
55128 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6AZZ1 | |
TRIM68 | |
Synthetic peptide directed towards the C-terminal of human TRIM68 (aa 397-446). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction