Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM68 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540894 Supplier Invitrogen™ Supplier No. PA540894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EPISLELKTD CRVLGLREIL KTYAADVRLD PDTAYSRLIV SEDRKRVHYG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 77%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 77%; Rat: 86%; Zebrafish: 83%.

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is expressed in many cancer cell lines. Its expression in normal tissues, however, was found to be restricted to prostate. This gene was also found to be differentially expressed in androgen-dependent versus androgen-independent prostate cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM68
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIM68
Gene Accession No. Q6AZZ1
Gene Alias AI894158; E3 ubiquitin-protein ligase TRIM68; F730114J12Rik; FLJ10369; GC109; LOC100720935; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM68; MGC126176; ring finger protein 137; RING-type E3 ubiquitin transferase TRIM68; RNF137; Ro/SSA1 related protein; SS56; SS-56; SSA protein SS-56; Trim68; tripartite motif containing 68; tripartite motif-containing 68; tripartite motif-containing protein 68
Gene Symbols TRIM68
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TRIM68.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55128
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.