Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM68 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582835
Description
Immunogen sequence: KLELNHSELI QQSQVLWRMI AELKERSQRP VRWMLQDIQE VLNRSKSWSL QQPEPISLEL KTDCRVLG.
The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is expressed in many cancer cell lines. Its expression in normal tissues, however, was found to be restricted to prostate. This gene was also found to be differentially expressed in androgen-dependent versus androgen-independent prostate cancer cells.
Specifications
TRIM68 | |
Polyclonal | |
Unconjugated | |
TRIM68 | |
AI894158; E3 ubiquitin-protein ligase TRIM68; F730114J12Rik; FLJ10369; GC109; LOC100720935; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM68; MGC126176; ring finger protein 137; RING-type E3 ubiquitin transferase TRIM68; RNF137; Ro/SSA1 related protein; SS56; SS-56; SSA protein SS-56; Trim68; tripartite motif containing 68; tripartite motif-containing 68; tripartite motif-containing protein 68 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55128 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6AZZ1 | |
TRIM68 | |
Recombinant protein corresponding to Human TRIM68. Recombinant protein control fragment (Product #RP-93614). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction