Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM71 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519281
Description
This antibody is predicted to react with bovine, canine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
TRIM71 is the mammalian ortholog to the C. elegans heterochronic protein Lin41, a protein that is thought to be important in postembryonic development, and is genetically and biochemically downstream of both Shh and Fgf signaling pathways. Similar to its C. elegans homolog, TRIM71 is a target of the microRNA let-7 and is likely to play a role in mammalian development. Recent experiments have indicated that TRIM71 is an E3 ubiquitin ligase and can interact with the Dicer and the Argonaute proteins Ago1, Ago2, and Ago4. Overexpression and depletion of TRIM71 led to inverse changes in Ago2 protein levels, suggesting TRIM71 can regulate Ago2 turnover. Finally, TRIM71 cooperates with the pluripotency factor Lin-28 in regulating let-7.
Specifications
| TRIM71 | |
| Polyclonal | |
| Unconjugated | |
| TRIM71 | |
| 2610206G21Rik; abnormal cell LINeage LIN-41; AL022943; B-box zinc finger, Filamin and NHL repeat containing protein (123.8 kD) (lin-41); Drosophila dappled/ vertebrate TRipartite Motif protein related; E3 ubiquitin-protein ligase TRIM71; Gm1127; homolog of C. elegans Lin-41; Lin41; LIN-41; lin-41 homolog; mLin41; mlin-41; protein lin-41 homolog; RGD1566388; RING-type E3 ubiquitin transferase TRIM71; Trim71; tripartite motif containing 71; tripartite motif containing 71, E3 ubiquitin protein ligase; tripartite motif-containing 71; tripartite motif-containing protein 71 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 131405, 636931 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q1PSW8, Q2Q1W2 | |
| TRIM71 | |
| Synthetic peptide sequence (RQVKAKSLYLQVE) corresponding to the internal amino acids of TRIM71 (aa 390-402). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction