Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143960
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143960 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143960 Supplier Invitrogen™ Supplier No. PA5143960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat skeletal muscle tissue, rat heart tissue, mouse skeletal muscle tissue, mouse heart tissue. IHC: human skeletal muscle tissue, mouse cardiac muscle tissue, rat heart tissue. Flow: THP-1 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Muscle-specific protein that plays a central role in cell membrane repair by nucleating the assembly of the repair machinery at injury sites. Specifically binds phosphatidylserine. Acts as a sensor of oxidation: upon membrane damage, entry of extracellular oxidative environment results in disulfide bond formation and homooligomerization at the injury site. This oligomerization acts as a nucleation site for recruitment of TRIM72-containing vesicles to the injury site, leading to membrane patch formation. Probably acts upstream of the Ca(2+)-dependent membrane resealing process. Required for transport of DYSF to sites of cell injury during repair patch formation. Regulates membrane budding and exocytosis. May be involved in the regulation of the mobility of KCNB1-containing endocytic vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM72
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene TRIM72
Gene Accession No. A0JPQ4, Q1XH17, Q6ZMU5
Gene Alias BC067209; mg53; mitsugumin 53; mitsugumin-53; RGD1562778; TRIM72; trim72 {ECO:0000250; tripartite motif containing 72; tripartite motif containing 72, E3 ubiquitin protein ligase; tripartite motif-containing 72; tripartite motif-containing protein 72; UniProtKB:Q8N4X6}
Gene Symbols TRIM72
Host Species Rabbit
Immunogen E.coli-derived human MG53/TRIM72 recombinant protein (Position: E28-A477).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 365377, 434246, 493829
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.