Learn More
Invitrogen™ TRIM74 Polyclonal Antibody

Description
The antibody detects endogenous levels of total TRIM74 protein.
Specifications
Specifications
| Antigen | TRIM74 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | TRIM74 |
| Gene Accession No. | Q86UV6 |
| Gene Alias | TRIM50C; TRIM74; tripartite motif containing 74; tripartite motif-containing 50C; tripartite motif-containing 74; tripartite motif-containing protein 50C; Tripartite motif-containing protein 74 |
| Gene Symbols | TRIM74 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.