Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531428 Supplier Invitrogen™ Supplier No. PA531428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, 3XFlag-human TRIM8-transfected 293T. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (91%), Cat (100%), Chicken (94%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to nuclear bodies. Its structure is similar to some tumor suppressor proteins and its gene maps to a locus thought to contain tumor suppressor genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TRIM8
Gene Accession No. Q9BZR9
Gene Alias AA408830; BB006755; E3 ubiquitin-protein ligase TRIM8; GERP; glioblastoma expressed ring finger protein; glioblastoma-expressed RING finger protein; Probable E3 ubiquitin-protein ligase TRIM8; ring finger protein 27; RING-type E3 ubiquitin transferase TRIM8; Rnf27; TRIM8; tripartite motif containing 8; tripartite motif protein 8; tripartite motif protein TRIM8; tripartite motif-containing 8; tripartite motif-containing protein 8; tripartite motif-containing protein 8; probable E3 ubiquitin-protein ligase TRIM8
Gene Symbols TRIM8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GERP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81603
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.