Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM9 Antibody (OTI2D6), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274616UV
Description
TRIM9 Monoclonal specifically detects TRIM9 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
TRIM9 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TRIM9 | |
Recombinant protein fragment corresponding to amino acids 284-669 of human TRIM9 (NP_443210) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI2D6 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
EC 6.3.2.-, KIAA0282homolog of rat RING finger Spring, RNF91RING finger protein 91, SPRINGE3 ubiquitin-protein ligase TRIM9, tripartite motif containing 9, tripartite motif-containing 9, Tripartite motif-containing protein 9 | |
Mouse | |
Affinity Purified | |
Zinc Finger | |
114088.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction