Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM9 Monoclonal Antibody (OTI2D6)
GREENER_CHOICE

Catalog No. PIMA526357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526357 Supplier Invitrogen™ Supplier No. MA526357
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRIM9 Monoclonal Antibody for Western Blot, ICC/IF, Flow

TRIM9 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM9
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TRIM9
Gene Accession No. Q9C026
Gene Alias E3 ubiquitin-protein ligase TRIM9; fj48c11; homolog of rat RING finger Spring; hypothetical protein LOC336099; Kiaa0282; RING finger protein 91; RING-type E3 ubiquitin transferase TRIM9; RNF91; SNAP-25-interacting RING finger protein; Spring; Trim9; tripartite motif containing 9; tripartite motif protein 9; tripartite motif-containing 9; tripartite motif-containing protein 9; tripartite motif-containing protein 9; E3 ubiquitin-protein ligase TRIM9; Unknown (protein for MGC:137652); wu:fj48c11; zgc:77357
Gene Symbols TRIM9
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 284-669 of TRIM9 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114088
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.