Learn More
TRIM9 Mouse anti-Human, Clone: OTI5G5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800067

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TRIM9 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms.The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms.Specifications
TRIM9 | |
Monoclonal | |
Unconjugated | |
TRIM9 | |
RNF91, SPRING | |
Mouse | |
Affinity Chromatography | |
RUO | |
114088 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5G5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9C026 | |
TRIM9 | |
Human recombit protein fragment corresponding to amino acids 284-669 of human TRIM9 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.