Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIM9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587152 Supplier Invitrogen™ Supplier No. PA587152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

TRIM9 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM9
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIM9
Gene Accession No. Q8C7M3, Q91ZY8, Q9C026
Gene Alias E3 ubiquitin-protein ligase TRIM9; fj48c11; homolog of rat RING finger Spring; hypothetical protein LOC336099; Kiaa0282; RING finger protein 91; RING-type E3 ubiquitin transferase TRIM9; RNF91; SNAP-25-interacting RING finger protein; Spring; Trim9; tripartite motif containing 9; tripartite motif protein 9; tripartite motif-containing 9; tripartite motif-containing protein 9; tripartite motif-containing protein 9; E3 ubiquitin-protein ligase TRIM9; Unknown (protein for MGC:137652); wu:fj48c11; zgc:77357
Gene Symbols TRIM9
Host Species Rabbit
Immunogen Recombinant fusion protein of human TRIM9.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114088, 155812, 94090
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.