Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoprim, Powder, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18607579 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 kg

T1537, 738-70-5, C14H18N4O3

Spectrum™ Chemical Trimethoprim, Powder, USP is an antibiotic used to treat bacterial infections. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 738-70-5
Molecular Formula C14H18N4O3
Molecular Weight (g/mol) 290.32
InChI Key IEDVJHCEMCRBQM-UHFFFAOYSA-N
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COC1=CC(CC2=CN=C(N)N=C2N)=CC(OC)=C1OC

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Fiber Drum
Quantity 25 kg
Percent Purity 98.5 to 101%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.