Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trimethoxyboroxine, Thermo Scientific™
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
Melting Point | 10°C to 11°C |
Color | Colorless |
Density | 1.2160g/mL |
Flash Point | 14°C |
Packaging | Glass bottle |
Refractive Index | 1.3996 |
Quantity | 25 g |
Specific Gravity | 1.216 |
Solubility Information | Solubility in water: reacts |
Viscosity | 21.7 mm2/s (20°C) |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement: Causes serious eye damage. Causes skin irritation. Highly flammable liquid and vapor. |
Precautionary Statement | GHS P Statement: Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 203-016-8 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction