Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl borate, 99.9995+% (metals basis)

Catalog No. p-7049809
Click to view available options
Quantity:
50 g
250 g

CAS: 121-43-7 | C3H9BO3 | 103.912 g/mol

Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in the Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-43-7
Molecular Formula C3H9BO3
Molecular Weight (g/mol) 103.912
MDL Number MFCD00008346
InChI Key WRECIMRULFAWHA-UHFFFAOYSA-N
Synonym trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
PubChem CID 8470
ChEBI CHEBI:38913
IUPAC Name trimethyl borate
SMILES B(OC)(OC)OC

Specifications

Melting Point -34°C
Color Colorless
Density 0.915 g/mL
Boiling Point 67°C to 69°C
Flash Point -8°C (17°F)
Odor Odorless
Assay Percent Range >99.9995% (metals basis)
Refractive Index 1.361
Linear Formula (CH3O)3B
Quantity 50 g
UN Number UN2416
Beilstein 1697939
Sensitivity Moisture sensitive
Merck Index 14,9712
Solubility Information Miscible with tetrahydrofuran,ether,isoporpylamine,hexane and methanol.
Formula Weight 103.92
Percent Purity ≥99.9995%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Trimethyl borate
Show More Show Less
Hazard Category H225-H312-H370
Hazard Statement GHS H Statement
H225-H312
Highly flammable liquid and vapor.
Harmful in contact with skin.
Precautionary Statement P210-P233-P240-P243-P260-P264b-P270-P280-P302+P352-P312-P314-P362-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIMETHYL BORATE
EINECSNumber 204-468-9
RTECSNumber ED5600000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.