Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl Phosphonoacetate 96.0+%, TCI America™
SDP

Catalog No. P1265250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1265250G 250 g
P126525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1265250G Supplier TCI America Supplier No. P1265250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5927-18-4
Molecular Formula C5H11O5P
Molecular Weight (g/mol) 182.112
MDL Number MFCD00008452
InChI Key SIGOIUCRXKUEIG-UHFFFAOYSA-N
Synonym trimethyl phosphonoacetate, methyl 2-dimethoxyphosphoryl acetate, trimethyl phosphonacetate, trimethylacetyl phosphate, acetic acid, dimethoxyphosphinyl-, methyl ester, methyl-dimethoxyphosphoryl acetat, trimethyl phosphoroacetate, phosphonoacetic acid trimethyl ester, methyl dimethylphosphonoacetate, methyl dimethoxyphosphoryl acetate
PubChem CID 80029
IUPAC Name methyl 2-dimethoxyphosphorylacetate
SMILES COC(=O)CP(=O)(OC)OC

Specifications

Color Colorless
Boiling Point 108°C
Quantity 250 g
Formula Weight 182.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Phosphonoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.