Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylacetaldehyde, 95%

Catalog No. aaa1501306
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 630-19-3 | C5H10O | 86.13 g/mol

Trimethylacetaldehyde is used as a stereoselective synthesis application. It is also used as a building block used in aldol condensation reactions. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetaldehyde is used as a stereoselective synthesis application. It is also used as a building block used in aldol condensation reactions. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is virtually insoluble in water but soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from oxidizing agents, strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-19-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00006962
InChI Key FJJYHTVHBVXEEQ-UHFFFAOYSA-N
Synonym trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde
PubChem CID 12417
IUPAC Name 2,2-dimethylpropanal
SMILES CC(C)(C)C=O

Specifications

Melting Point 6°C
Density 0.775
Boiling Point 74°C to 76°C
Flash Point −15°C (5°F)
Odor Pleasant
Refractive Index 1.3794
Linear Formula (CH3)3CCHO
Quantity 5 g
UN Number UN1989
Beilstein 506060
Sensitivity Air sensitive
Solubility Information It is virtually insoluble in water but soluble in chloroform.
Formula Weight 86.13
Percent Purity 96%
Chemical Name or Material Trimethylacetaldehyde
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 211-134-6
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.