Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylacetonitrile, 98%

Catalog No. p-3737666
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle

CAS: 630-18-2 | C5H9N | 83.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-18-2
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00001847
InChI Key JAMNHZBIQDNHMM-UHFFFAOYSA-N
Synonym pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn
PubChem CID 12416
IUPAC Name 2,2-dimethylpropanenitrile
SMILES CC(C)(C)C#N

Specifications

Melting Point 15°C to 16°C
Color Colorless
Density 0.7500g/mL
Boiling Point 105°C to 106°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3765 to 1.3785
Linear Formula (CH3)3CCN
Quantity 25 mL
Beilstein 02, 320
Specific Gravity 0.75
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone and toluene
Formula Weight 83.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Trimethylacetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF
EINECSNumber 211-133-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.