Alfa Aesar™ Trimethylamine, 1M soln. in THF

Manufacturer: Alfa Aesar™ H32899AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH32899AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-50-3
    Molecular Formula C3H9N
    Linear Formula (CH3)3N
    Formula Weight 59.11
    MDL Number MFCD00008327
    UN Number UN1993
    Chemical Name or Material Trimethylamine

    Specifications

    Formula Weight 59.11
    UN Number UN1993
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Concentration 1M soln. in THF
    Quantity 500mL
    TSCA TSCA
    Molecular Formula C3H9N
    Linear Formula (CH3)3N
    MDL Number MFCD00008327
    CAS 75-50-3
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H224-H318-H302-H332-H335-H315
    Extremely flammable liquid and vapor.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause respiratory irritation.
    Causes skin irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    RTECS Number PA0350000
    Chemical Name or Material Trimethylamine

    Description

    Applications
    Trimethylamine complexes of alane as precursors for the low-pressure chemical vapor deposition of aluminum. Trimethylamine was found to be unsuitable as an agent for differentiating between Lewis and Brpinsted sites because it is dissociatively adsorbed and is, therefore useful for the perturbing effects. Choline derivative manufactured from TMA are used in medication of liver disorders. Used directly as acid binder in the manufacture of Sulpha Drugs. Animal/poultry feed additive like choline chloride is manufactured from TMA. Used in the manufacture of alkyl quaternary ammonium salts. To manufacture Ion exchange Resins based on chloromethylated polystyrene.

    Solubility
    Easily soluble in water, also soluble in most organic solvents (such as methanol and ethanol).

    Notes
    Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

    GHS H Statement
    H224-H318-H302-H332-H335-H315
    Extremely flammable liquid and vapor.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit