Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol

Catalog No. AC270440250
Click to view available options
Quantity:
25 mL
250 mL
1 L
5 L

Catalog No. AC270440250

Supplier: Thermo Scientific Chemicals 270440250

Only null left
Add to Cart

CAS: 75-50-3 | C3H9N | 59.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethylamine
CAS 75-50-3, 64-17-5
Color Colorless
Density 0.7500g/mL
Boiling Point 50.0°C
Flash Point −20°C
Packaging Glass bottle
Molecular Formula C3H9N
Refractive Index 1.3630 to 1.3660
Linear Formula (CH3)3N
Quantity 25 mL
MDL Number MFCD00008327
Beilstein 04,43
Merck Index 15,988
Specific Gravity 0.75
Synonym trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,benzene,toluene,,xylene,ethylbenzene and chloroform
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
SMILES CN(C)C
IUPAC Name trimethylamine
Molecular Weight (g/mol) 59.11
PubChem CID 1146
ChEBI CHEBI:18139
Formula Weight 59.11
Percent Purity 32 to 34%
Grade Pure
Physical Form Liquid
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.