Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylboroxine

Catalog No. 90396525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
5 g
250 mL
Danger

Catalog No. 90-396-525ML

Supplier: Sigma Aldrich 90396525ML

Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylboroxine (TMB) is a cyclic anhydride of methyl-boronic acid. It can be used as a:Methylating agent for the methylation of various aromatic halides and C(sp3)-H bonds using palladium catalyst.Reagent in the preparation of polymer supported CBS (Corey, Bakshi, and Shibata) catalysts.Organoboranes. 39. Convenient procedures for the preparation of methylboronic acid and trimethylboroxin

Specifications

CAS 823-96-1
Density 0.89962 g/mL
Molecular Formula C3H9B3O3
Linear Formula C3H9B3O3
Refractive Index n/D 1.3880
MDL Number MFCD00013354
Quantity 250 mL
Molecular Weight (g/mol) 125.53 g/mol
Biological Activity Respiratory System
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.