Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylboroxine, 3.5M (50 wt.%) solution in THF, AcroSeal™

Catalog No. ac429191000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 823-96-1 | C3H9B3O3 | 125.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823-96-1, 109-99-9
Molecular Formula C3H9B3O3
Molecular Weight (g/mol) 125.53
MDL Number MFCD00013354
InChI Key GBBSAMQTQCPOBF-UHFFFAOYSA-N
Synonym trimethylboroxine, 2,4,6-trimethylboroxin, trimethylboroxin, boroxin, trimethyl, trimethyl-1,3,5,2,4,6-trioxatriborinane, trimethyboroxine, trimethyl boroxin, trimethylboroxaine, trimethyl boroxine, trimethyl-boroxine
PubChem CID 574072
IUPAC Name 2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane
SMILES CB1OB(C)OB(C)O1

Specifications

Melting Point -38.0°C
Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 78.0°C to 80.0°C
Flash Point −9°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.89
Formula Weight 125.54
Percent Purity 40.0 to 60.0% (ex Boron)
Physical Form Solution
Chemical Name or Material Trimethylboroxine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Suspected of causing cancer.
May cause drowsi
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.