Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylene Di(thiotosylate) 95.0+%, TCI America™
SDP

Catalog No. D239025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D239025G 25 g
D23905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D239025G Supplier TCI America Supplier No. D239025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3866-79-3
Molecular Formula C17H20O4S4
Molecular Weight (g/mol) 416.583
MDL Number MFCD00014918
InChI Key QOICHLMIPNOKLN-UHFFFAOYSA-N
Synonym 1,3-Di(p-tosylthio)propane, 1,3-Propanediyl Di(thiotosylate), Trimethylene Bis(p-toluenethiosulfonate)
PubChem CID 256129
IUPAC Name 1-methyl-4-[3-(4-methylphenyl)sulfonylsulfanylpropylsulfanylsulfonyl]benzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)SCCCSS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 25 g
Formula Weight 416.58
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylene Di(thiotosylate) [Protecting Reagent for Active Methylene]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.