Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylhydroquinone, 97%

Catalog No. p-207030
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

CAS: 700-13-0 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-13-0
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002346
InChI Key AUFZRCJENRSRLY-UHFFFAOYSA-N
Synonym trimethylhydroquinone, 2,3,5-trimethylhydroquinone, 1,4-benzenediol, 2,3,5-trimethyl, pseudocumohydroquinone, psi-cumohydroquinone, 2,3,6-trimethylhydroquinone, 2,3,5-trimethyl-benzene-1,4-diol, hydroquinone, trimethyl, 2,3,5-trimethyl-1,4-benzenediol, .psi.-cumohydroquinone
PubChem CID 12785
IUPAC Name 2,3,5-trimethylbenzene-1,4-diol
SMILES CC1=CC(=C(C(=C1O)C)C)O

Specifications

Melting Point 169°C to 174°C
Color Beige
Boiling Point 295°C
Flash Point 191°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 06, IV, 5997
Solubility Information Solubility in water: 2g/L water (20°C). Other solubilities: soluble in most common organic solvents
Formula Weight 152.19
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Trimethylhydroquinone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
May cause an allergic skin reaction.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiou
EINECSNumber 211-838-3
RTECSNumber MX7820000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.