Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylolpropane triacrylate, stabilized

Catalog No. p-7049624
Click to view available options
Quantity:
100 g
500 g
2 kg

CAS: 15625-89-5 | C15H20O6 | 296.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15625-89-5
Molecular Formula C15H20O6
Molecular Weight (g/mol) 296.32
MDL Number MFCD00008628
InChI Key DAKWPKUUDNSNPN-UHFFFAOYSA-N
Synonym trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s
PubChem CID 27423
ChEBI CHEBI:35028
IUPAC Name 2,2-bis(prop-2-enoyloxymethyl)butyl prop-2-enoate
SMILES CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C

Specifications

Density 1.10 g/mL
Boiling Point >200°C (1 mmHg)
Flash Point 87°C (188°F)
Odor Pungent
Refractive Index 1.4735
Quantity 100 g
Solubility Information Miscible with water.
Formula Weight 296.31
Physical Form Liquid
Chemical Name or Material Trimethylolpropane triacrylate, Stabilized
Hazard Category H227-H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H317-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c
EINECSNumber 239-701-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.