Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Trimethylsilyl)acetylene, 98%

Catalog No. p-7020785
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1066-54-2 | C5H10Si | 98.22 g/mol

(Trimethylsilyl)acetylene is used in the preparation of trimethylsilanyl-propiolic acid ethyl ester by reacting with carbonochloridic acid ethyl ester as well as in the synthesis of iodoalkenes by radical addition of perfluoroalkyl iodides. It is used as a nucleophile in Friedel-Crafts type acylations and alkylations reactions, as a ligand in organometallic chemistry and an useful reagent in cycloaddition reactions. It acts as a precursor to lithium trimethylsilylacetylide It finds application in the synthesis of (±)-estrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Trimethylsilyl)acetylene is used in the preparation of trimethylsilanyl-propiolic acid ethyl ester by reacting with carbonochloridic acid ethyl ester as well as in the synthesis of iodoalkenes by radical addition of perfluoroalkyl iodides. It is used as a nucleophile in Friedel-Crafts type acylations and alkylations reactions, as a ligand in organometallic chemistry and an useful reagent in cycloaddition reactions. It acts as a precursor to lithium trimethylsilylacetylide It finds application in the synthesis of (±)-estrone.

Solubility
Miscible with organic solvents.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-54-2
Molecular Formula C5H10Si
Molecular Weight (g/mol) 98.22
MDL Number MFCD00008569
InChI Key CWMFRHBXRUITQE-UHFFFAOYSA-N
Synonym trimethylsilylacetylene, trimethylsilyl acetylene, silane, ethynyltrimethyl, ethynyl-trimethyl-silane, ethynyl trimethyl silane, tms acetylene, ethynyltrimethyl silane, trimethylsilyl-acetylene, tmsacetylene
PubChem CID 66111
IUPAC Name ethynyl(trimethyl)silane
SMILES C[Si](C)(C)C#C

Specifications

Boiling Point 52°C to 53°C
Quantity 5 g
Linear Formula HC≡CSi(CH3)3
UN Number UN1993
Beilstein 906752
Flash Point <−34°C (−29°F)
Solubility Information Miscible with organic solvents.
Formula Weight 98.22
Percent Purity 98%
Refractive Index 1.39
Density 0.71
Chemical Name or Material (Trimethylsilyl)acetylene
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 213-919-9
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.