Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilyl cyanide, 97%

Catalog No. AAA1959814
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 7677-24-9 | C4H9NSi | 99.21 g/mol

Trimethylsilyl Cyanide is used as a reagent in the preparation of optically active cyanohydrins. It is used in the derivatization of complex metabolite mixtures by GC-MS. It is employed in the preparation of Reissert compounds, which represent reactive polyamides. Further, it is used to convert pyridine-N-oxides into 2-cyanopyridine. It is also used as a cyanide source in an enantioselective organocatalytic Strecker-type reaction of aliphatic N,N-dialkylhydrazones. In addition to this, it is used as a reagent for the cyanosilylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl Cyanide is used as a reagent in the preparation of optically active cyanohydrins. It is used in the derivatization of complex metabolite mixtures by GC-MS. It is employed in the preparation of Reissert compounds, which represent reactive polyamides. Further, it is used to convert pyridine-N-oxides into 2-cyanopyridine. It is also used as a cyanide source in an enantioselective organocatalytic Strecker-type reaction of aliphatic N,N-dialkylhydrazones. In addition to this, it is used as a reagent for the cyanosilylation of aldehydes.

Solubility
Miscible with organic solvents.

Notes
Moisture sensitive. Store in a cool place.Incompatible with water, strong oxidizing agents, strong reducing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7677-24-9
Molecular Formula C4H9NSi
Molecular Weight (g/mol) 99.21
MDL Number MFCD00001765
InChI Key LEIMLDGFXIOXMT-UHFFFAOYSA-N
Synonym trimethylsilyl cyanide, cyanotrimethylsilane, trimethylsilanecarbonitrile, trimethylsilylcyanide, silanecarbonitrile, trimethyl, trimethylsilylnitrile, tmscn, trimethyl silane cyanide, trimethylsilylcarbonitrile, tms cyanide
PubChem CID 82115
IUPAC Name trimethylsilylformonitrile
SMILES C[Si](C)(C)C#N

Specifications

Boiling Point 118°C to 119°C
Melting Point 11°C to 12°C
Quantity 25 g
UN Number UN3383
Beilstein 1737612
Flash Point 1°C (33°F)
Solubility Information Miscible with organic solvents.
Formula Weight 99.21
Percent Purity 97%
Odor Almond-like
Refractive Index 1.391
Sensitivity Moisture sensitive
Density 0.783
Chemical Name or Material Trimethylsilyl cyanide
Show More Show Less
Hazard Category H225-H300+H310+H330
Hazard Statement GHS H Statement
H225-H300-H310-H330
Highly flammable liquid and vapour.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
EINECSNumber 231-657-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.