Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trimethylsilyl cyanide, 98%
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent

Specifications
Specifications
Boiling Point | 114.0°C to 117.0°C |
Melting Point | 11.0°C |
Quantity | 500 g |
Assay Percent Range | 98% |
Linear Formula | (CH3)3SiCN |
Flash Point | 1°C |
Fieser | 04,542; 05,720; 06,632; 07,397; 08,133; 09,127; 10,112; 11,147; 12,148; 13,87; 14,107; 15,102; 17,89; 01,979; 04,418; 06,503; 08,433 |
Solubility Information | Solubility in water: reacts. |
Formula Weight | 99.21 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement: Fatal if swallowed. Fatal if inhaled. Very toxic to aquatic life with long lasting effects. Fatal in contact with skin. Highly flammable liquid and vapor. Contact with water liberates toxic gas. |
Precautionary Statement | GHS P Statement: Avoid release to the environment. Wear protective gloves/protective clothing/eye protection/face protection. Do not breathe dust/fume/gas/mist/vapors/spray. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction