Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilyl Isocyanate 95.0+%, TCI America™
SDP

Catalog No. T110625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T110625G 25 g
T11065G 5 g
T1106100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T110625G Supplier TCI America Supplier No. T110625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1118-02-1
Molecular Formula C4H9NOSi
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001993
InChI Key NIZHERJWXFHGGU-UHFFFAOYSA-N
Synonym trimethylsilyl isocyanate, silane, isocyanatotrimethyl, trimethylsilylisocyanate, isocyanato trimethyl silane, trimethylisocyanatosilane, tmsisocyanate, tms isocyanate, tms-isocyanate, tms isocynate
PubChem CID 70696
IUPAC Name isocyanatotrimethylsilane
SMILES C[Si](C)(C)N=C=O

Specifications

Color Colorless
Boiling Point 90°C
Quantity 25 g
UN Number 2478
Formula Weight 115.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.