Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilyl trifluoromethanesulfonate, 99%

Catalog No. AAA1253518
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1253518 50 g
AAA1253509 10 g
AAA1253530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1253518 Supplier Thermo Scientific Chemicals Supplier No. A1253518
Add to Cart

CAS: 27607-77-8 | C4H9F3O3SSi | 222.253 g/mol

Trimethylsilyl trifluoromethanesulfonate is used to prepare powerful Lewis acid, difluoroboron triflate etherate in acetonitrile solvent. It is also used as a reagent in a Dieckmann-like cyclization of ester-imides and diesters. Further, it is used in the conversion of carbonyl compounds to their enol ethers. It is also employed for chemical glycosylation reactions. Its reactivity is similar to trimethylsilyl chloride and is also involved in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl trifluoromethanesulfonate is used to prepare powerful Lewis acid, difluoroboron triflate etherate in acetonitrile solvent. It is also used as a reagent in a Dieckmann-like cyclization of ester-imides and diesters. Further, it is used in the conversion of carbonyl compounds to their enol ethers. It is also employed for chemical glycosylation reactions. Its reactivity is similar to trimethylsilyl chloride and is also involved in organic synthesis.

Solubility
Miscible with aliphatic, aromatic hydrocarbons, haloalkanes and ethers. Immiscible with water.

Notes
Moisture and air sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27607-77-8
Molecular Formula C4H9F3O3SSi
Molecular Weight (g/mol) 222.253
MDL Number MFCD00000406
InChI Key FTVLMFQEYACZNP-UHFFFAOYSA-N
Synonym trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate
PubChem CID 65367
IUPAC Name trimethylsilyl trifluoromethanesulfonate
SMILES C[Si](C)(C)OS(=O)(=O)C(F)(F)F

Specifications

Density 1.225
Boiling Point 39°C to 40°C (12 mmHg)
Flash Point 25°C (77°F)
Refractive Index 1.36
Linear Formula (CH3)3SiSO3CF3
Quantity 50 g
UN Number UN2924
Beilstein 1868911
Sensitivity Moisture sensitive
Merck Index 14,9719
Solubility Information Miscible with aliphatic,aromatic hydrocarbons,haloalkanes and ethers. Immiscible with water.
Formula Weight 222.26
Percent Purity 99%
Chemical Name or Material Trimethylsilyl trifluoromethanesulfonate
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 248-565-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.