Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilylmethanethiol 97.0+%, TCI America™
SDP

Catalog No. T12131ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 mL

Chemical Identifiers

CAS 18165-76-9
Molecular Formula C4H12SSi
Molecular Weight (g/mol) 120.285
MDL Number MFCD00059112
InChI Key IIYTXTQVIDHSSL-UHFFFAOYSA-N
Synonym Trimethylsilylmethyl Mercaptan
PubChem CID 140342
IUPAC Name trimethylsilylmethanethiol
SMILES C[Si](C)(C)CS

Specifications

Color Colorless
Boiling Point 116°C
Quantity 1 mL
UN Number 1993
Formula Weight 120.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilylmethanethiol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.