Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilylmethyl Azide 97.0+%, TCI America™
SDP

Catalog No. T11841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11841G 1 g
T11845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11841G Supplier TCI America Supplier No. T11841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 87576-94-1
Molecular Formula C4H11N3Si
Molecular Weight (g/mol) 129.24
MDL Number MFCD00059964
InChI Key HKBUFTCADGLKAS-UHFFFAOYSA-N
Synonym Azidomethyltrimethylsilane
PubChem CID 150751
IUPAC Name (azidomethyl)trimethylsilane
SMILES C[Si](C)(C)CN=[N+]=[N-]

Specifications

Boiling Point 60°C
Quantity 1 g
UN Number 1993
Formula Weight 129.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilylmethyl Azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.