Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylstannylbutyldimethylsilane, 97%, Thermo Scientific™

Catalog No. AC382500010
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103731-29-9
Molecular Formula C9H24SiSn
Molecular Weight (g/mol) 279.086
MDL Number MFCD03093828
InChI Key CIBQAISGPNTXQG-UHFFFAOYSA-N
Synonym butyl tert-butyl methylsilyl ? 3-stannyl
PubChem CID 57346340
IUPAC Name butyl-tert-butyl-methylsilicon;$l^{3}-stannane
SMILES CCCC[Si](C)C(C)(C)C.[SnH3]

Specifications

Color Colorless
Boiling Point 101.0°C to 103.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Refractive Index 1.4820 to 1.4840 (20°C, 589nm)
Linear Formula (CH3)3SnSi(CH3)2(CH2)3CH3
Quantity 1 g
Formula Weight 279.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Trimethylstannylbutyldimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if swallowed.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.