Learn More
Trimethylsulfonium hydroxide, 0.25M solution in methanol, ACROS Organics™
Specifications
Concentration | 0.2 to 0.3M |
Density | 0.81g/mL |
Formula Weight | 94.18 |
Physical Form | Liquid |
Packaging | Glass bottle |
Color | Colorless |
Quantity | 10mL |
Chemical Name or Material | Trimethylsulfonium hydroxide, 0.25M solution in methanol |
Safety and Handling
GHS H Statement:
Causes damage to organs.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger