Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsulfoxonium Iodide 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
1774-47-6
C3H9IOS
220.07
MFCD00011899
BPLKQGGAXWRFOE-UHFFFAOYSA-M
trimethylsulfoxonium iodide,trimethylsulphoxonium iodide,trimethyloxosulfonium iodide,sulfoxonium, trimethyl-, iodide,trimethyloxosulphonium iodide,trimethyl sulphoxonium ioidie,s,s,s-trimethylsulfoxonium iodide,trimethylsulfoxoniumiodide,sulfonium, trimethyl-, iodide, oxide,sulfoxonium, trimethyl-, iodide 1:1
74498
C[S+](=O)(C)C.[I-]
1774-47-6
C3H9IOS
220.07
MFCD00011899
BPLKQGGAXWRFOE-UHFFFAOYSA-M
trimethylsulfoxonium iodide,trimethylsulphoxonium iodide,trimethyloxosulfonium iodide,sulfoxonium, trimethyl-, iodide,trimethyloxosulphonium iodide,trimethyl sulphoxonium ioidie,s,s,s-trimethylsulfoxonium iodide,trimethylsulfoxoniumiodide,sulfonium, trimethyl-, iodide, oxide,sulfoxonium, trimethyl-, iodide 1:1
74498
C[S+](=O)(C)C.[I-]
Color White to Yellow
Quantity 500g
Infrared Spectrum Authentic
Melting Point 175.0°C
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula (CH3)3S(O)I
Solubility Information Solubility in water: 15g/L (20°C). Other solubilities: insoluble in ethanol and ether
Formula Weight 220.07
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Trimethylsulfoxonium iodide, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 217-204-2

RTECSNumber : WS3585000

TSCA : TSCA