Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIML1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569235 Supplier Invitrogen™ Supplier No. PA569235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Probable E3 ubiquitin-protein ligase which plays an important role in blastocyst development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIML1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Triml1
Gene Accession No. Q8N9V2
Gene Alias 4933403D14; BC050188; probable E3 ubiquitin-protein ligase TRIML1; RGD1305337; RING finger protein 209; RING-type E3 ubiquitin transferase TRIML1; RNF209; Triml1; tripartite motif family like 1; tripartite motif family-like 1; tripartite motif family-like protein 1
Gene Symbols Triml1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TRIML1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339976
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.