Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trinity Biotech Buffers for Premier Hb9210 Analyzers
Click to view available options
Type:
Premier™ Buffer A
Premier™ Buffer B
Description
- Performance validated to assure accuracy and precision
- Ready for use
- 940mL
Specifications
Specifications
| Content And Storage | 15°C to 28°C |
| Type | Premier™ Buffer A |
| For Use With (Application) | Quantitative measurement of hemoglobin A1c, in-vitro diagnostic use only |
| Quantity | 940 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction