Learn More
Epredia™ Cassette 1 Biopsy Cassettes
Click to view available options
Color:
Blue
Gray
Green
Lilac
Pink
White
Yellow
Description
- Made of acetal polymer
- Cassette writing face is at a 30° angle
- Pore style is small square
- Locking lid attached by two hinges
- Supplied in a box of 500 cassettes with attached breakaway lids
Order Info
Not available in Tube Packs
Specifications
Specifications
Pore Style | Square |
Writing Face Angle | 30° |
Closure Type | Hinged |
Color | White |
Material | Acetal Polymer |
Product Type | Biopsy Cassette |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov