Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543590 Supplier Invitrogen™ Supplier No. PA543590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PGFVLGHTSA VIVENPDGTL KKSTSWHTAL RLRKKSEKKD KDGKREGKLE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

TRIO promotes the exchange of GDP by GTP. Together with leukocyte antigen-related (LAR) protein, it could play a role in coordinating cell-matrix and cytoskeletal rearrangements necessary for cell migration and cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIO
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIO
Gene Accession No. O75962
Gene Alias 6720464I07Rik; AA408740; ARHGEF23; LOW QUALITY PROTEIN: triple functional domain protein; PTPRF-interacting protein; si:ch211-13c6.1; si:dkey-158b13.2; Solo; tgat; trio; trio Rho guanine nucleotide exchange factor; trio Rho guanine nucleotide exchange factor a; trioa; triple functional domain (PTPRF interacting); triple functional domain protein
Gene Symbols TRIO
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TRIO (aa 2607-2656).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7204
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.