Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552520
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552520 Supplier Invitrogen™ Supplier No. PA552520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLVTHDYTAV KEDEINVYQG EVVQILASNQ QNMFLVFRAA TDQCPAAEGW IPGFVLGHTS AVIVENPDGT LKKSTSWHTA LRLRKKSEKK DKDGKREGKL ENGYRKSREG LSNKVSVKLL NPN Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

TRIO promotes the exchange of GDP by GTP. Together with leukocyte antigen-related (LAR) protein, it could play a role in coordinating cell-matrix and cytoskeletal rearrangements necessary for cell migration and cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIO
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIO
Gene Accession No. O75962
Gene Alias 6720464I07Rik; AA408740; ARHGEF23; LOW QUALITY PROTEIN: triple functional domain protein; PTPRF-interacting protein; si:ch211-13c6.1; si:dkey-158b13.2; Solo; tgat; trio; trio Rho guanine nucleotide exchange factor; trio Rho guanine nucleotide exchange factor a; trioa; triple functional domain (PTPRF interacting); triple functional domain protein
Gene Symbols TRIO
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIO. Recombinant protein control fragment (Product #RP-89828).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7204
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.