Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIO Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580188
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL.
TRIO promotes the exchange of GDP by GTP. Together with leukocyte antigen-related (LAR) protein, it could play a role in coordinating cell-matrix and cytoskeletal rearrangements necessary for cell migration and cell growth.
Specifications
TRIO | |
Polyclonal | |
Unconjugated | |
TRIO | |
6720464I07Rik; AA408740; ARHGEF23; LOW QUALITY PROTEIN: triple functional domain protein; PTPRF-interacting protein; si:ch211-13c6.1; si:dkey-158b13.2; Solo; tgat; trio; trio Rho guanine nucleotide exchange factor; trio Rho guanine nucleotide exchange factor a; trioa; triple functional domain (PTPRF interacting); triple functional domain protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
223435, 310192, 7204 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
F1M0Z1, O75962, Q0KL02 | |
TRIO | |
A synthetic peptide corresponding to a sequence at the C-terminus of human TRIO (787-830aa QLRIFERDAIDIISDLESWNDELSQQMNDFDTEDLTIAEQRLQH). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction