Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIO, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABN145225UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN145225UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN145225UG Supplier MilliporeSigma Supplier No. ABN145225UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-TRIO Antibody, Cat. No. ABN1452, is a highly specific rabbit polyclonal antibody that targets Triple functional domain protein (TRIO) and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.

Triple functional domain protein (UniProt: O75962; also known as EC: 2.7.11.1, PTPRF-interacting protein, TRIO) is encoded by the TRIO gene (Gene ID: 7204) in human. TRIO is a complex protein that is widely expressed in human tissue, with highest levels reported in heart, skeletal muscle, and brain. Five isoforms of TRIO have been reported that are produced by alternative splicing. It contains three catalytic domains, two tandem Dbl Homology (DH) and pleckstrin homology (PH) domains, and a serine/Threonine kinase domain. The DH-PH module is involved in guanine nucleotide exchange and the activation of the small GTPases of the Rho family. The DH1-PH1 domain (GEF1) is reported to activate the Rho G and Rac1, whereas DH2-PH2 (GEF2) is involved in the activation of RhoA. TRIO is involved in coordinating actin remodeling, which is necessary for cell migration and growth. In developing hippocampal neurons it limits dendrite formation, without affecting the establishment of axon polarity. After dendrites are formed, it is reported to be involved in the control of synaptic function by regulating the endocytosis of AMPA-selective glutamate receptors at CA1 excitatory synapses. Mutations in TRIO gene have been linked to mental retardation and microcephaly. (Ref.: Debant, A. et al. (1996). Proc. Natl. Acad. Sci, USA. 93(11); 5466-71; Bellanger, JM. Et al. (2000). Nat. Cell Biol. 2(12); 888-92).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIO
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected TRIO in human cerebral cortex, human pancreas, and human testis tissue.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Triple functional domain protein;EC: 2.7.11.1.
Gene Symbols TRIO
Host Species Rabbit
Immunogen KLH-conjugated liner peptide corresponding to 17 amino acids from the C-terminal half of human Triple functional domain protein.
Molecular Weight of Antigen ∽300 kDa observed; 346.90 kDa calculalted. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects human Triple functional domain protein (TRIO). It targets an epitope within 17 amino acids from the C-terminal half. The immunogen sequence is conserved in all 5 isoforms.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.