Alfa Aesar™ Trioctyl phosphate

Manufacturer: Alfa Aesar™ 15808A1

Quantity 1kg

 View more versions of this product

Catalog No. AA15808A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78-42-2
    Formula Weight 434.64
    MDL Number MFCD00009491
    Synonym Tris(2-ethylhexyl) phosphate
    Chemical Name or Material Trioctyl phosphate

    Specifications

    Formula Weight 434.64
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P362-P312
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Take off contaminated clothing and wash before reuse.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Refractive Index 1.4410
    Physical Form Liquid
    CAS 78-42-2
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H332-H315-H319
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    TSCA TSCA
    MDL Number MFCD00009491
    Synonym Tris(2-ethylhexyl) phosphate
    Density 0.93g/mL
    Flash Point >110° (230°F)
    Boiling Point 216°C (4mmHg)
    Quantity 1kg
    Chemical Name or Material Trioctyl phosphate

    GHS H Statement
    H332-H315-H319
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P362-P312
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Take off contaminated clothing and wash before reuse.
    Call a POISON CENTER or doctor/physician if you feel unwell.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit