Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trioctylphosphine

Catalog No. 718165100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
100 mL
500 mL
2 L
Danger

Catalog No. 71-816-5100ML

Supplier: Sigma Aldrich 718165100ML

Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trioctylphosphine can be used for the conversion of metal nanocrystals, bulk powders, foils, wires, thin films to metal phosphides. It can also act as a solvent and stabilizer for synthesizing cadmium sulfide nanorods from cadmium acetate and sulfur.

Specifications

CAS 4731-53-7
Density 0.831 g/mL at 25°C
Boiling Point 284°C to 291°C (50 mmHg, lit.)
Molecular Formula C24H51P
Refractive Index n20/D 1.468
Linear Formula [CH3(CH2)7]3P
Quantity 100 mL
MDL Number MFCD00015298
Synonym TOP; P(Oct)3
Molecular Weight (g/mol) 370.64 g/mol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.