Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triosephosphate isomerase Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB300994
Description
Triosephosphate isomerase Polyclonal antibody specifically detects Triosephosphate isomerase in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA.Specifications
Triosephosphate isomerase | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
EC 5.3.1.1, MGC88108, TIM, TPI, triosephosphate isomerase, Triose-phosphate isomerase, triosephosphate isomerase 1 | |
Goat | |
26.7 kDa | |
0.1 mg | |
Primary | |
This antibody is expected to recognise reported isoforms 1, 2 and 3 (NP_000356.1; NP_001152759.1, NP_001244955.1) | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.001 - 0.01 μg/mL, Flow Cytometry 10 μg/mL, Immunohistochemistry 2 - 4 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL, Peptide ELISA Detection limit 1:16000 | |
P60174 | |
TPI1 | |
Peptide with sequence C-LKPEFVDIINAKQ corresponding to C-Terminus according to NP_000356.1; NP_001152759.1; NP_001244955.1. | |
Affinity Purified | |
RUO | |
7167 | |
Human, Mouse, Rat, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction