Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triosephosphate isomerase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131470
Description
Triosephosphate isomerase Polyclonal antibody specifically detects Triosephosphate isomerase in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
Triosephosphate isomerase | |
Polyclonal | |
Unconjugated | |
1XPBS, 1% BSA, 10% Glycerol (pH7). with 0.025% Proclin 300 | |
EC 5.3.1.1, MGC88108, TIM, TPI, triosephosphate isomerase, Triose-phosphate isomerase, triosephosphate isomerase 1 | |
Rabbit | |
31 kDa | |
100 μL | |
Stem Cell Markers | |
7167 | |
Human, Mouse, Rat, Bovine | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
P60174 | |
TPI1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Triosephosphate isomerase. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Predicted cross-reactivity based on sequence identoty: Yeast (83%), Xenopus tropicalis (92%), and Chimpanzee (100%). Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction